Assay Details
| Gene Symbol: ZBTB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076264.2 | NP_001069732.1 | 3 - 4 | 1071 | 67 | |
| GenBank mRNA | BC120135.1 | - | 3 - 4 | 1073 | 67 | |
More Information
Set Membership: |
|

ZBTB8A
Genomic Map