Assay Details
| Gene Symbol: ZBTB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076264.2 | NP_001069732.1 | 4 - 5 | 1244 | 78 | |
| GenBank mRNA | BC120135.1 | - | 4 - 5 | 1246 | 78 | |
More Information
Set Membership: |
|

ZBTB8A
Genomic Map