Assay Details
| Gene Symbol: ICE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076265.1 | NP_001069733.1 | 11 - 12 | 2539 | 92 | |
| GenBank mRNA | BC120051.1 | - | 11 - 12 | 2539 | 92 | |
More Information
Set Membership: |
|

ICE2
Genomic Map