Assay Details
| Gene Symbol: FAM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076309.1 | NP_001069777.1 | 3 - 4 | 275 | 58 | |
| GenBank mRNA | BC120368.1 | - | 3 - 4 | 275 | 58 | |
More Information
Set Membership: |
|

FAM3A
Genomic Map