Assay Details
| Gene Symbol: GALNT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076316.1 | NP_001069784.1 | 10 - 11 | 1861 | 69 | |
| GenBank mRNA | BC122597.1 | - | 10 - 11 | 1861 | 69 | |
More Information
Set Membership: |
|

GALNT18
Genomic Map