Assay Details
| Gene Symbol: MAP6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076354.1 | NP_001069822.1 | 1 - 2 | 382 | 150 | |
| GenBank mRNA | BC120336.1 | - | 1 - 2 | 382 | 150 | |
More Information
Set Membership: |
|

MAP6D1
Genomic Map