Assay Details
| Gene Symbol: TMEM39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076360.2 | NP_001069828.1 | 7 - 8 | 1462 | 64 | |
| GenBank mRNA | BC120195.1 | - | 7 - 8 | 1463 | 64 | |
| BT030725.1 | - | 7 - 8 | 1465 | 64 | ||
More Information
Set Membership: |
|

TMEM39A
Genomic Map