Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076367.1 | NP_001069835.1 | 1 - 2 | 103 | 127 | |
| GenBank mRNA | BC119888.1 | - | 1 - 2 | 103 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC119888.1, NM_001076367.1 transcripts.
Set Membership: |
|

ORMDL3
Genomic Map