Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076367.1 | NP_001069835.1 | 2 - 3 | 299 | 64 | |
| GenBank mRNA | BC119888.1 | - | 2 - 3 | 299 | 64 | |
More Information
Set Membership: |
|

ORMDL3
Genomic Map