Assay Details
| Gene Symbol: SEPTIN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076371.1 | NP_001069839.1 | 6 - 7 | 563 | 75 | |
| GenBank mRNA | BC120325.1 | - | 6 - 7 | 563 | 75 | |
More Information
Set Membership: |
|

SEPTIN5
Genomic Map