Assay Details
| Gene Symbol: EIF1AD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076393.1 | NP_001069861.1 | 5 - 6 | 747 | 86 | |
| GenBank mRNA | BC120113.1 | - | 5 - 6 | 747 | 86 | |
| BT021815.1 | - | 5 - 6 | 86 | |||
More Information
Set Membership: |
|

EIF1AD
Genomic Map