Assay Details
| Gene Symbol: NRADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076398.1 | NP_001069866.1 | 2 - 3 | 261 | 158 | |
| GenBank mRNA | BC105517.1 | - | 2 - 3 | 286 | 158 | |
More Information
Set Membership: |
|

NRADD
Genomic Map