Assay Details
| Gene Symbol: PHYHIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076415.1 | NP_001069883.1 | 1 - 2 | 218 | 77 | |
| GenBank mRNA | BC119858.1 | - | 1 - 2 | 248 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC119858.1, NM_001076415.1 transcripts.
Set Membership: |
|

PHYHIP
Genomic Map