Assay Details
| Gene Symbol: PHYHIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076415.1 | NP_001069883.1 | 4 - 5 | 706 | 66 | |
| GenBank mRNA | BC119858.1 | - | 4 - 5 | 736 | 66 | |
More Information
Set Membership: |
|

PHYHIP
Genomic Map