Assay Details
| Gene Symbol: CAPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076424.1 | NP_001069892.1 | 3 - 4 | 604 | 67 | |
| GenBank mRNA | BC120244.1 | - | 3 - 4 | 604 | 67 | |
More Information
Set Membership: |
|

CAPS
Genomic Map