Assay Details
| Gene Symbol: RRAGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076456.1 | NP_001069924.1 | 4 - 5 | 848 | 82 | |
| GenBank mRNA | BC119864.1 | - | 4 - 5 | 848 | 82 | |
More Information
Set Membership: |
|

RRAGC
Genomic Map