Assay Details
| Gene Symbol: SPATA2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076502.2 | NP_001069970.1 | 1 - 2 | 59 | 59 | |
| GenBank mRNA | BC122735.1 | - | 1 - 2 | 60 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122735.1, NM_001076502.2 transcripts.
Set Membership: |
|

SPATA2L
Genomic Map