Assay Details
| Gene Symbol: SYBU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076511.2 | NP_001069979.1 | 4 - 5 | 588 | 72 | |
| GenBank mRNA | BC119820.1 | - | 4 - 5 | 589 | 72 | |
| BC147975.1 | - | 8 - 9 | 1074 | 72 | ||
More Information
Set Membership: |
|

SYBU
Genomic Map