Assay Details
| Gene Symbol: HMGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076523.1 | NP_001069991.1 | 4 - 5 | 443 | 142 | |
| GenBank mRNA | BC120383.1 | - | 4 - 5 | 443 | 142 | |
More Information
Set Membership: |
|

HMGA1
Genomic Map