Assay Details
| Gene Symbol: ANGPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076797.1 | NP_001070265.1 | 3 - 4 | 912 | 147 | |
| GenBank mRNA | AF093573.1 | - | 3 - 4 | 721 | 147 | |
| BC123628.1 | - | 3 - 4 | 912 | 147 | ||
More Information
Set Membership: |
|

ANGPT1
Genomic Map