Assay Details
| Gene Symbol: BMPR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076800.1 | NP_001070268.1 | 10 - 11 | 1574 | 62 | |
| GenBank mRNA | AJ550059.1 | - | 1 - 2 | 160 | 62 | |
| DQ922946.1 | - | 10 - 11 | 1574 | 62 | ||
More Information
Set Membership: |
|

BMPR1A
Genomic Map