Assay Details
| Gene Symbol: RABL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076817.1 | NP_001070285.1 | 3 - 4 | 482 | 61 | |
| GenBank mRNA | BC123865.1 | - | 3 - 4 | 482 | 61 | |
More Information
Set Membership: |
|

RABL6
Genomic Map