Assay Details
| Gene Symbol: TAGAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076823.1 | NP_001070291.1 | 4 - 5 | 296 | 76 | |
| GenBank mRNA | BC123848.1 | - | 4 - 5 | 296 | 76 | |
More Information
Set Membership: |
|

TAGAP
Genomic Map