Assay Details
| Gene Symbol: RWDD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076824.1 | NP_001070292.1 | 4 - 5 | 562 | 143 | |
| GenBank mRNA | BC123494.1 | - | 4 - 5 | 562 | 143 | |
More Information
Set Membership: |
|

RWDD4
Genomic Map