Assay Details
| Gene Symbol: PARP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076828.1 | NP_001070296.1 | 8 - 9 | 2152 | 74 | |
| GenBank mRNA | BC123641.1 | - | 8 - 9 | 2152 | 74 | |
More Information
Set Membership: |
|

PARP9
Genomic Map