Assay Details
| Gene Symbol: SH3GL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076840.2 | NP_001070308.1 | 7 - 8 | 865 | 56 | |
| GenBank mRNA | BC123532.1 | - | 7 - 8 | 866 | 56 | |
More Information
Set Membership: |
|

SH3GL2
Genomic Map