Assay Details
| Gene Symbol: CELF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076845.1 | NP_001070313.1 | 11 - 12 | 1593 | 58 | |
| GenBank mRNA | BC123480.1 | - | 11 - 12 | 1593 | 58 | |
| BC149370.1 | - | 7 - 8 | 791 | 58 | ||
| BT029831.1 | - | 9 - 10 | 1281 | 58 | ||
More Information
Set Membership: |
|

CELF3
Genomic Map