Assay Details
| Gene Symbol: UGCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076850.1 | NP_001070318.1 | 8 - 9 | 1129 | 92 | |
| GenBank mRNA | BC123602.1 | - | 8 - 9 | 1129 | 92 | |
More Information
Set Membership: |
|

UGCG
Genomic Map