Assay Details
| Gene Symbol: PRKCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076863.1 | NP_001070331.1 | 11 - 12 | 1613 | 52 | |
| GenBank mRNA | BC120027.1 | - | 11 - 12 | 1613 | 52 | |
More Information
Set Membership: |
|

PRKCH
Genomic Map