Assay Details
| Gene Symbol: CFAP100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076866.1 | NP_001070334.1 | 15 - 16 | 1881 | 53 | |
| GenBank mRNA | BC123870.1 | - | 15 - 16 | 1881 | 53 | |
More Information
Set Membership: |
|

CFAP100
Genomic Map