Assay Details
| Gene Symbol: CFAP100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076866.1 | NP_001070334.1 | 1 - 2 | 59 | 72 | |
| GenBank mRNA | BC123870.1 | - | 1 - 2 | 59 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC123870.1, NM_001076866.1 transcripts.
Set Membership: |
|

CFAP100
Genomic Map