Assay Details
| Gene Symbol: CAMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076868.1 | NP_001070336.1 | 5 - 6 | 547 | 69 | |
| GenBank mRNA | BC123625.1 | - | 5 - 6 | 547 | 69 | |
More Information
Set Membership: |
|

CAMK1
Genomic Map