Assay Details
| Gene Symbol: BRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076873.1 | NP_001070341.1 | 10 - 11 | 2095 | 79 | |
| GenBank mRNA | BC123578.1 | - | 10 - 11 | 2095 | 79 | |
More Information
Set Membership: |
|

BRD3
Genomic Map