Assay Details
| Gene Symbol: CSDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076877.1 | NP_001070345.1 | 3 - 4 | 568 | 66 | |
| GenBank mRNA | BC123465.1 | - | 3 - 4 | 568 | 66 | |
More Information
Set Membership: |
|

CSDC2
Genomic Map