Assay Details
| Gene Symbol: RB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076907.1 | NP_001070375.1 | 4 - 5 | 676 | 69 | |
| GenBank mRNA | BC123388.1 | - | 4 - 5 | 676 | 69 | |
More Information
Set Membership: |
|

RB1
Genomic Map