Assay Details
| Gene Symbol: PPIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076910.1 | NP_001070378.1 | 1 - 2 | 187 | 148 | |
| GenBank mRNA | BC123474.1 | - | 1 - 2 | 187 | 148 | |
More Information
Set Membership: |
|

PPIC
Genomic Map