Assay Details
| Gene Symbol: PPIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076910.1 | NP_001070378.1 | 2 - 3 | 295 | 82 | |
| GenBank mRNA | BC123474.1 | - | 2 - 3 | 295 | 82 | |
More Information
Set Membership: |
|

PPIC
Genomic Map