Assay Details
| Gene Symbol: SCN4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076912.1 | NP_001070380.1 | 4 - 5 | 715 | 107 | |
| GenBank mRNA | BC123479.1 | - | 4 - 5 | 715 | 107 | |
More Information
Set Membership: |
|

SCN4B
Genomic Map