Assay Details
| Gene Symbol: WIPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076923.2 | NP_001070391.1 | 4 - 5 | 544 | 65 | |
| GenBank mRNA | BC123527.1 | - | 4 - 5 | 545 | 65 | |
More Information
Set Membership: |
|

WIPF1
Genomic Map