Assay Details
| Gene Symbol: SMAD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076928.1 | NP_001070396.1 | 2 - 3 | 865 | 138 | |
| GenBank mRNA | DQ922948.1 | - | 2 - 3 | 865 | 138 | |
More Information
Set Membership: |
|

SMAD9
Genomic Map