Assay Details
| Gene Symbol: RGS7BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076930.1 | NP_001070398.1 | 1 - 2 | 639 | 53 | |
| GenBank mRNA | BC123745.1 | - | 1 - 2 | 639 | 53 | |
More Information
Set Membership: |
|

RGS7BP
Genomic Map