Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076949.2 | NP_001070417.1 | 1 - 2 | 127 | 81 | |
| GenBank mRNA | BC123664.1 | - | 1 - 2 | 109 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC123664.1, NM_001076949.2 transcripts.
Set Membership: |
|

SEPTIN3
Genomic Map