Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076949.2 | NP_001070417.1 | 2 - 3 | 321 | 70 | |
| GenBank mRNA | BC123664.1 | - | 2 - 3 | 303 | 70 | |
More Information
Set Membership: |
|

SEPTIN3
Genomic Map