Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076949.2 | NP_001070417.1 | 9 - 10 | 1036 | 69 | |
| GenBank mRNA | BC123664.1 | - | 9 - 10 | 1018 | 69 | |
More Information
Set Membership: |
|

SEPTIN3
Genomic Map