Assay Details
| Gene Symbol: UBALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076951.1 | NP_001070419.1 | 2 - 3 | 259 | 107 | |
| GenBank mRNA | BC123814.1 | - | 2 - 3 | 259 | 107 | |
More Information
Set Membership: |
|

UBALD1
Genomic Map