Assay Details
| Gene Symbol: B3GALNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076963.1 | NP_001070431.1 | 3 - 4 | 172 | 146 | |
| GenBank mRNA | BC120120.1 | - | 3 - 4 | 172 | 146 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120120.1, NM_001076963.1 transcripts.
Set Membership: |
|

B3GALNT1
Genomic Map