Assay Details
| Gene Symbol: TMCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077135.1 | NP_001070603.1 | 1 - 2 | 661 | 66 | |
| GenBank mRNA | BC123417.1 | - | 1 - 2 | 661 | 66 | |
More Information
Set Membership: |
|

TMCC2
Genomic Map