Assay Details
| Gene Symbol: CTSZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077835.1 | NP_001071303.1 | 5 - 6 | 867 | 81 | |
| GenBank mRNA | BC122603.1 | - | 5 - 6 | 867 | 81 | |
| X01809.1 | - | 117 | 81 | |||
More Information
Set Membership: |
|

CTSZ
Genomic Map