Assay Details
| Gene Symbol: CASP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077840.1 | NP_001071308.1 | 4 - 5 | 400 | 119 | |
| GenBank mRNA | AY575000.1 | - | 2 - 3 | 179 | 119 | |
| BC123503.1 | - | 4 - 5 | 400 | 119 | ||
More Information
Set Membership: |
|

CASP3
Genomic Map