Assay Details
| Gene Symbol: TXNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077886.2 | NP_001071354.1 | 4 - 5 | 664 | 77 | |
| GenBank mRNA | BC122816.1 | - | 4 - 5 | 665 | 77 | |
More Information
Set Membership: |
|

TXNL1
Genomic Map